Learn More
Hydrazine sulfate, ACS, 99.0% min
CAS: 10034-93-2 | H6N2O4S | 130.118 g/mol
Supplier: Thermo Scientific Chemicals 04012036
Description
Reagent for determination of C-terminal amino acidsHydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hydrazine sulfate | |
| 10034-93-2 | |
| 500 g | |
| H6N2O4S | |
| hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| hydrazine;sulfuric acid | |
| 24842 | |
| ACS Reagent |
| ∽255°C (decomposition) | |
| Crystalline | |
| 99.0% min | |
| MFCD00044873 | |
| Soluble in water. Insoluble in alcohol. | |
| NN.OS(=O)(=O)O | |
| 130.118 | |
| 130.12 | |
| 1.37 g/mL |
Chemical Identifiers
| 10034-93-2 | |
| 130.118 | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| 24842 | |
| NN.OS(=O)(=O)O |
| H6N2O4S | |
| MFCD00044873 | |
| hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
| hydrazine;sulfuric acid |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H350
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 233-110-4
RTECSNumber : MV9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only