missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Monohydrochloride 98.0+%, TCI America™

Catalog No. H017425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H017425G 25 g
H0174500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H017425G Supplier TCI America Supplier No. H017425G
Only null left

Chemical Identifiers

CAS 2644-70-4
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.504
MDL Number MFCD00044368
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422
PubChem CID 115031
IUPAC Name hydrazine;hydrochloride
SMILES NN.Cl

Specifications

Melting Point 93°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3290
Formula Weight 68.50
Percent Purity ≥98.0% (T)
Chemical Name or Material Hydrazine Monohydrochloride
EINECSNumber (1)-0374
RTECSNumber MV4625000
TSCA Yes