missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Monohydrobromide 98.0+%, TCI America™

Catalog No. H017325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H017325G 25 g
H0173500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H017325G Supplier TCI America Supplier No. H017325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity crystalline powder for laboratory research applications

  • Laboratory grade reagent used for research and analytical applications
  • Solid laboratory reagent to be stored under inert gas in a cool and dark place <15°C

Chemical Identifiers

CAS 13775-80-9
Molecular Formula BrH5N2
Molecular Weight (g/mol) 112.958
MDL Number MFCD00065394
InChI Key DVHXJLRODLTJOD-UHFFFAOYSA-N
Synonym hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
PubChem CID 159693
IUPAC Name hydrazine;hydrobromide
SMILES NN.Br

Specifications

Melting Point 88°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range >98.0%
UN Number 3290
Solubility Information Soluble in Water
Formula Weight 112.96
Percent Purity ≥98.0% (T)
Sensitivity Light Sensitive, Air Sensitive, Hygroscopic
Appearance White to Almost White Powder to Crystal
Chemical Name or Material Hydrazine Monohydrobromide
Show More Show Less