missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Monohydrobromide (ca. 50% in Water), TCI America™

Catalog No. H151725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H151725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H151725G Supplier TCI America Supplier No. H151725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13775-80-9
Molecular Formula BrH5N2
Molecular Weight (g/mol) 112.958
MDL Number MFCD00065394
InChI Key DVHXJLRODLTJOD-UHFFFAOYSA-N
Synonym hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
PubChem CID 159693
IUPAC Name hydrazine;hydrobromide
SMILES NN.Br

Specifications

Color Colorless
Quantity 25 g
UN Number 3289
Formula Weight 112.96
Physical Form Liquid
Chemical Name or Material Hydrazine Monohydrobromide (ca. 50% in Water)