missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Monohydrate 98.0+%, TCI America™

Catalog No. p-7130383
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H017225ML 25 mL
H0172500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H017225ML Supplier TCI America Supplier No. H017225ML
Only null left

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H6N2O
Molecular Weight (g/mol) 50.06
MDL Number MFCD00149931
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine hydrate
SMILES O.NN

Specifications

Boiling Point 119°C
Melting Point -52°C
Color Colorless
Physical Form Liquid
Quantity 25 mL
UN Number 2030
Formula Weight 32.05
Percent Purity ≥98.0% (T)
Chemical Name or Material Hydrazine Monohydrate
EINECSNumber (1)-0374
RTECSNumber MV8050000
TSCA Yes