missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Monohydrate (79%), TCI America™

Catalog No. p-7130403
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H020425G 25 g
H0204500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H020425G Supplier TCI America Supplier No. H020425G
Only null left

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H6N2O
Molecular Weight (g/mol) 50.06
MDL Number MFCD00149931
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine hydrate
SMILES O.NN

Specifications

Melting Point -52°C
Color Colorless
Boiling Point 119°C
Quantity 25 g
UN Number 2030
Formula Weight 32.05
Physical Form Liquid
Chemical Name or Material Hydrazine Monohydrate (79%)
EINECSNumber (1)-0374
RTECSNumber MV8050000
TSCA No