missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Hydrate, Technical, 85%, Spectrum™ Chemical

Catalog No. p-7165631
Change view
Click to view available options
Quantity:
10 mL
250 mL
1 L
5 gal.
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610824 10 mL Amber Glass Bottle
18610825 250 mL Amber Glass Bottle
18610829 1 L Amber Glass Bottle
18610826 5 gal. Poly Drum
18610827 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610824 Supplier Spectrum Chemical Mfg Cor Supplier No. H101410ML
Only null left

Chemical Identifiers

CAS 7803-57-8

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 10 mL
Percent Purity 0.85
Grade Technical

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.