missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Dihydrobromide Hydrate 98.0+%, TCI America™

Catalog No. H016925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H016925G 25 g
H0169500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H016925G Supplier TCI America Supplier No. H016925G

Chemical Identifiers

CAS 23268-00-0
Molecular Formula Br2H6N2
Molecular Weight (g/mol) 193.87
MDL Number MFCD00070601
InChI Key MEBDASBFCISMGU-UHFFFAOYSA-N
Synonym Hydrazinium Dibromide
PubChem CID 168095
IUPAC Name dihydrogen hydrazine dibromide
SMILES [H+].[H+].[Br-].[Br-].NN

Specifications

Color White
pH 4.5 to 7.5 (300g/L H2O 20°C)
Boiling Point 178°C
Quantity 25 g
UN Number 3290
Formula Weight 193.87
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hydrazine Dihydrobromide Hydrate
EINECSNumber (1)-0105&(1)-0374
TSCA Yes