missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Carbonate (70% in Water, ca. 7.3mol/L), TCI America™

Catalog No. H085425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H085425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H085425G Supplier TCI America Supplier No. H085425G
Only null left

Chemical Identifiers

CAS 112077-84-6
Molecular Formula CH6N2O3
Molecular Weight (g/mol) 94.07
MDL Number MFCD00185569
InChI Key PTYMQUSHTAONGW-UHFFFAOYSA-N
PubChem CID 14693823
IUPAC Name carbonic acid;hydrazine
SMILES C(=O)(O)O.NN

Specifications

Color Colorless
pH 9.4
Quantity 25 g
Formula Weight 108.1
Physical Form Liquid
Chemical Name or Material Hydrazine Carbonate (70% in Water, ca. 7.3mol/L)