Learn More
Eisco™ Hydraulic Press
Catalog No. S24880
Change view
Click to view available options
Hydraulic Press
Catalog No.
S24880 Hydraulic Press
1 options
Catalog No. S24880 Supplier Eisco™ Supplier No. PH0215
For demonstrating transmissibility of fluid pressure.
  • Shows how mechanical advantage is obtained using interconnected cylinders of different diameters
  • Graduated glass syringes with capacities of 10 and 50mL and cross-sectional area of 3 to 1
  • Pistons provided with loading platform of 90mm dia.
Specifications
Product Type Hydraulic Press
For Use With (Application) Demonstrating the transmissibility of fluid pressure
Includes Graduated syringe barrels of 10 mL and 50 mL, pistons with loading platforms 90 mm in diameter