missing translation for 'onlineSavingsMsg'
Learn More

Whirl-Pak™ Hydrated PolySponge™ Sampling Bag with Sampling Sponge
GREENER_CHOICE

Catalog No. 14209180
Change view
Click to view available options
Capacity (Metric):
532 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14209180 532 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14209180 Supplier Whirl-Pak™ Supplier No. B01590
Only null left

Makes environmental surface sampling faster and easier.

  • Specially designed for the Food & Beverage industry
  • Write-on label makes identifying samples easy and convenient
  • Bags are pre-hydrated with HiCap™ Neutralizing Broth
  • HiCap Broth provides superior neutralization, validated up to 72 hr.
  • Polyurethane sponges included with each bag are non toxic and do not contain biocides
  • Each box contains five foil pouches with 20 sponges each
  • Terminally gamma processed to guarantee sterility
  • Medical-grade, super absorbent, and ideal for rough surfaces
  • Compatible with 3M Petrifilm
  • 2 year shelf life

Specifications

Description Without Glove
Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
For Use With (Application) Environmental Monitoring
Material Polyurethane
Sterility Sterile
Fill Volume 16 mL
Latex Free Yes
No. per Case 100/Cs
No. per Pack 100/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Foil Pouch
Sterilization Method Terminally Gamma Processed
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 2.50 mil
Length (English) 9 in.
Length (English) Document Pouch 9 in.
Length (Metric) Document Pouch 23 cm
Width (English) 4.5 in.
Width (English) Document Pouch 4 in.
Width (Metric) Document Pouch 11.5 cm
Product Type Sample Bag with Sampling Sponge
Capacity (English) 18 oz.
Capacity (Metric) 532 mL
Length (Metric) 23 cm
Width (Metric) 11.5 cm
Thickness (Metric) 0.064 mm
Show More Show Less