missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG, Anolyte for coulometric Karl Fischer titration (methanol-based), for cells with and without diaphragm, Fluka, Honeywell

Catalog No. 60049048
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60049048 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60049048 Supplier Honeywell Inc Supplier No. 348361LUS
Only null left

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)

Chemical Identifiers

Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

Boiling Point 63°C
CAS Min % ≥5.00, ≥70.00
CAS Max % <10.00
CAS 68007-08-9, 7446-09-5, 67-56-1, 111-42-2, 288-32-4
Density 0.930 g/cm3
Physical Form Liquid
Quantity 1 L
Flash Point 14°C
Packaging Polycoated Bottle
Grade Coulometry
Hazard Category Danger
Hazard Statement H373+H225+H318+H332+H360+H370
Precautionary Statement P280+P271+P270+P264+P260+P243+P201+P202+P210+P233+P240+P242+P241
Recommended Storage Room Temperature