missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solstice Advanced HYDRANAL-COMPOSITE 5 K, 2.5LT
Supplier: Honeywell Inc 348164X25LUS
HYDRANAL-COMPOSITE 5 K, 2.5LT
Specifications
| ∼1.15Â g/mL | |
| 4 x 2.5L | |
| Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free) | |
| ≥70.0000%,≥5.0000%,≥5.0000%,≥1.0000%,≥1.0000%,≥1.0000% | |
| Glass Bottle | |
| Karl Fischer |
| 99°C | |
| HYDRANAL™ - Composite 5 K | |
| 111-90-0,7553-56-2,288-32-4,693-98-1,7446-09-5,68007-08-9 | |
| <90.0000%,<10.0000%,<10.0000%,<5.0000%,<5.0000%,<5.0000% | |
| DE Solvent, 2-(2-Ethoxyethoxy)ethanol, CARBITOL™, Diethylene glycol ethyl ether, Ethyldiglycol |