missing translation for 'onlineSavingsMsg'
Learn More

Hydantoin 99.0+%, TCI America™

Catalog No. H0167500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0167500G 500 g
H016725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0167500G Supplier TCI America Supplier No. H0167500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 461-72-3
Molecular Formula C3H4N2O2
Molecular Weight (g/mol) 100.08
MDL Number MFCD00005259
InChI Key WJRBRSLFGCUECM-UHFFFAOYSA-N
Synonym hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol
PubChem CID 10006
ChEBI CHEBI:27612
IUPAC Name imidazolidine-2,4-dione
SMILES O=C1CNC(=O)N1

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 500 g
Formula Weight 100.08
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hydantoin
EINECSNumber (9)-1026
RTECSNumber MT8210000
TSCA Yes