missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydantoin, 99%
CAS: 461-72-3 | C3H4N2O2 | 100.08 g/mol
$62.51 - $723.78
Chemical Identifiers
| CAS | 461-72-3 |
|---|---|
| Molecular Formula | C3H4N2O2 |
| Molecular Weight (g/mol) | 100.08 |
| MDL Number | MFCD00005259 |
| InChI Key | WJRBRSLFGCUECM-UHFFFAOYSA-N |
| Synonym | hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol |
| PubChem CID | 10006 |
| ChEBI | CHEBI:27612 |
| IUPAC Name | imidazolidine-2,4-dione |
| SMILES | O=C1CNC(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1248622
|
Thermo Scientific Chemicals
A1248622 |
100 g |
Each for $62.51
|
|
|||||
|
AAA1248636
|
Thermo Scientific Chemicals
A1248636 |
500 g |
Each for $190.66
|
|
|||||
|
AAA124860E
|
Thermo Scientific Chemicals
A124860E |
2500 g |
Each for $723.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 461-72-3 | |
| 100.08 | |
| WJRBRSLFGCUECM-UHFFFAOYSA-N | |
| 10006 | |
| imidazolidine-2,4-dione |
| C3H4N2O2 | |
| MFCD00005259 | |
| hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol | |
| CHEBI:27612 | |
| O=C1CNC(=O)N1 |
Specifications
| 461-72-3 | |
| C3H4N2O2 | |
| 100 g | |
| 14,4761 | |
| WJRBRSLFGCUECM-UHFFFAOYSA-N | |
| imidazolidine-2,4-dione | |
| 10006 | |
| 100.08 | |
| Hydantoin |
| 221°C to 223°C | |
| MFCD00005259 | |
| 110598 | |
| hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol | |
| O=C1CNC(=O)N1 | |
| 100.08 | |
| CHEBI:27612 | |
| 99% |
Safety and Handling
P262-P314-P501c
H373
EINECSNumber : 207-313-3
RTECSNumber : MT8210000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only