missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydantoic Acid 98.0+%, TCI America™
Supplier: TCI America H065525G
Specifications
| Hydantoic Acid | |
| White | |
| MFCD00047876 | |
| hydantoic acid, n-carbamoylglycine, carbamoylglycine, glycoluric acid, n-carbamylglycine, glycine, n-aminocarbonyl, acetic acid, ureido, urea, carboxymethyl, 2-ureidoacetic acid, carbomoylglycine | |
| C(C(=O)O)NC(=O)N | |
| 118.092 | |
| 118.09 | |
| Crystalline Powder |
| 462-60-2 | |
| C3H6N2O3 | |
| 25 g | |
| KZVRXPPUJQRGFN-UHFFFAOYSA-N | |
| 2-(carbamoylamino)acetic acid | |
| 10020 | |
| ≥98.0% (T) |
Chemical Identifiers
| 462-60-2 | |
| 118.092 | |
| KZVRXPPUJQRGFN-UHFFFAOYSA-N | |
| 10020 | |
| C(C(=O)O)NC(=O)N |
| C3H6N2O3 | |
| MFCD00047876 | |
| hydantoic acid, n-carbamoylglycine, carbamoylglycine, glycoluric acid, n-carbamylglycine, glycine, n-aminocarbonyl, acetic acid, ureido, urea, carboxymethyl, 2-ureidoacetic acid, carbomoylglycine | |
| 2-(carbamoylamino)acetic acid |
Safety and Handling
RTECSNumber : MB7660000
TSCA : Yes