missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ RPMI 1640 Media

Designed to support cell culture growth and biomanufacturing processes.

Supplier:  Cytiva SH30096.LS

 View more versions of this product

Catalog No. SH30096LS


Only null left

Description

Description

  • Format: Liquid
Specifications

Chemical Identifiers

Liquid
0.1 μm Sterile-filtered
2° to 8°C, away from light
RPMI 1640
ISO 9001:2000

Specifications

Liquid
0.1 μm Sterile-filtered
No Glutamine
ISO 9001:2000
RPMI 1640
Phenol Red
6 x 1000 mL
2° to 8°C, away from light
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 Months

SDS
Documents

Documents

Promotions

Promotions