missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HyClone™ Peak Expression Medium

Catalog No. SH3119302
Change view
Click to view available options
Reconstituted Volume:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reconstituted Volume
SH3119302 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3119302 Supplier Cytiva™ Supplier No. SH31193.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use medium supports growth and efficient transfection of adeno-associated virus (AAV) expression in HEK 293 cell lines.

This medium supports high cell growth, high transfection efficiency, and robust viral production for a variety of AAV serotypes expressed in human embryonic kidney (HEK) cell lines. HyClone™ peak expression medium is an animal-derived-component free (ADCF), hydrolysate-free and regulatory-friendly cell culture medium. The versatility of this medium allows high transfection efficiency, consistent cell growth, high viable cell density, and high viral titers across many HEK 293 cell lines and AAV serotypes.

HyClone™ peak expression medium supports a seamless transition from growth to production of AAV using standard cell lines and processes. HyClone™ peak expression medium is available in liquid and powder formats in user-friendly packaging.

  • Ready-to-use medium contains stable L-glutamine and poloxamer 188
  • Designed for high cell yield and robust virus production
  • Allows for direct adaptation
  • Designed for small- to large-scale transfection and production applications
  • Compatible with a variety of PEI- and lipid-based transfection reagents

Specifications

Cell Line HEK
Cell Type Embryonic Kidney
Classification Viral Production
Form Liquid
Product Type Cell Culture Media
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Poloxamer 188
Without Additives None
Reconstituted Volume 1 L
Culture Type Mammalian Cell Culture
Expression System Mammalian
Show More Show Less