missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Insect Cell Culture Media
Increase cell yield with multiple insect lines in a variety of culture environments with HyClone™ Insect Cell Culture Media.
Supplier: Cytiva SH3006502
Specifications
| Liquid | |
| Glutamine | |
| 1000Â mL | |
| PETE |
| 0.1 μm Sterile-filtered | |
| No Sodium Bicarbonate, No Phenol Red, No HEPES | |
| 2° to 8°C, away from light |