missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ HyCell TransFx-H Powdered Media

Catalog No. SH3094401
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3094401 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3094401 Supplier Cytiva Supplier No. SH30944.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Animal-derived component-free (ADCF), hydrolysate-free, and regulatory-friendly cell culture medium.

The versatility of this medium allows quick adaptation and supports exceptional transfection, high viable cell density, and high productivity across a variety of human embryonic kidney (HEK) 293 cell lines.

HyCell TransFx-H is available in liquid and powder formats in user friendly packaging.

  • Hydrolysate-free and ADCF formulation
  • Supports high transfection efficiency
  • Designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Designed for micro- to large-scale transfection and production applications
  • Manufactured from traceable components according to cGMP guidelines

Specifications

Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No Poloxamer 188
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture