missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4MAb™ Media
Description
Increase process yields for the manufacture of monoclonal antibodies for therapeutic use with HyClone™ CDM4MAb™ Media, a chemically-defined media containing no animal derived components.
Specifications
Specifications
| Form | Liquid |
| Product Type | CDM4MAb Liquid Medium |
| Sterilization Method | 0.2 μm Sterile-filtered |
| With Additives | Sodium Bicarbonate |
| Without Additives | No Glutamine, No Poloxamer 188, No Phenol Red, No HEPES |
| Quantity | 1000 mL bottle |
| Content And Storage | 2° to 8°C, away from light |
| Description | Without L-Glutamine |