missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ CDM4Avian Powdered Media
GREENER_CHOICE

Catalog No. SH3103501
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3103501 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3103501 Supplier Cytiva Supplier No. SH31035.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for vaccine manufacturing using suspension cell culture with avian cell line.

HyClone CDM4Avian cell culture medium is developed for manufacturing of viral vaccines in avian cell lines, such as EB66 cells, and for the production of lytic viruses, such as measles, influenza, and Japanese encephalitis (JE); and of non-lytic viruses, such as the modified vaccinia Ankara (MVA) virus.

  • Chemically defined (CD) animal-derived component-free (ADCF) formulation
  • Designed to support high peak cell density as well as enhanced viral infectivity and productivity
  • Economical, operation-reducing single formulation for cell propagation and viral production
  • Designed for small- to large-scale culture applications
  • Available in liquid and powder formulations

Specifications

Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No Phenol Red, No Sodium Bicarbonate, No Poloxamer 188
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture