missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Amphotericin B (Fungiezone) Solution

Catalog No. SV3007801
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SV3007801 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SV3007801 Supplier Cytiva Supplier No. SV30078.01
Only null left
  • An antibiotic agent that is active against fungi
  • Used as an antifungal, anti-yeast and anti-mold agent
  • Interferes with the permeability of the cell membrane of sensitive fungi by binding sterols
  • Common working concentration is 2.5μg/mL, and it is cytotoxic at 30μg/mL

Specifications

Content And Storage Store frozen
Description Amphotericin B (Fungizone) Solution, 250μg/mL
Antibiotic Spectrum Antimycotic, Antifungal
Concentration 250 μg/mL
Form Liquid
Quantity 50 mL