Learn More
Cytiva HyClone™ AdvanceSTEM™ Neural Differentiation Kit and Growth Supplement
Catalog No. SH3087802
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
SH3087802 50 mL
1 options
Catalog No. SH3087802 Supplier Cytiva Supplier No. SH30878.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Kit includes a proprietary basal Neural Differentiation Medium to which an aliquot of AdvanceSTEM Growth Supplement (SH3087802) is added at time of use. Basal Neural Differentiation Medium and Growth Supplement should be used together, but they are also available as separate components.
Developed to facilitate the optimal neural differentiation of Adipose-derived Mesenchymal Stem Cells (HAMSC), Bone Marrow Mesenchymal Stem Cells (HBMSC)and Multipotent Cord Blood Unrestricted Somatic Stem Cells (MCBUSSCs)
Specifications
Product Type Stem Cell Differentiation Media
For Use With (Application) Neural Differentiation of Adipose-derived Mesenchymal Stem Cells (HAMSC), Bone Marrow Mesenchymal Stem Cells (HBMSC)and Multipotent Cord Blood Unrestricted Somatic Stem Cells (MCBUSSCs)
Quantity 50 mL