missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ AdvanceSTEM™ Cell Culture Media

Catalog No. SH3082401
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3082401 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3082401 Supplier Cytiva Supplier No. SH30824.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Support the growth of embryonic cell cultures with HyClone™ AdvanceSTEM™ Cell Culture Media.

Specifications

Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) 
Form Liquid
Product Type Stem Cell Media
Sterility Sterile-filtered
With Additives Glucose
Without Additives No Glutamine, No Sodium Pyruvate
Quantity 500 mL
Content And Storage Store at 2° to 8°C away from light
Description AdvanceSTEM DMEM4SC with 4500mg/L Glucose, without L-glutamine and Sodium Pyruvate