missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ 1 M NaOH - ADCF

Catalog No. p-200004033
Change view
Click to view available options
Quantity:
1 L
10 L
15 L
50 L
90 L
200 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3108801HY 1 L
SH3108802 10 L
SH3108803 15 L
SH3108804 50 L
SH3108805 90 L
SH3108806 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SH3108801HY Supplier Cytiva Supplier No. SH31088.01
Only null left

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean your chromatography resins.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use.
  • Not for diagnostic or therapeutic use.
  • 0.1 μm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.
  • HyClone™ NaOH in single-use bags is well-suited for use with: Cytiva's chromatography resins.
  • Available in 0.1 M (0.1 N), 0.5 M (0.5 N), and 1.0 M (1.0 N) sodium hydroxide concentrations up to 200 L.

Specifications

Chemical Name or Material Sodium Hydroxide
pH >12.0 at 20 to 25°C
Color Clear
Molecular Formula NaOH
Concentration 1.0 M (1.0 N)
Quantity 1 L
Grade NF/USP
Packaging Single-use Bag