missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Hybridization Tubes
GREENER_CHOICE

Catalog No. 1138116B
Change view
Click to view available options
Length (Metric):
150 mm
300 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
1138116B 150 mm
1138116C 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1138116B Supplier Corning™ Supplier No. 7995150

Includes

Plug seal cap with O-ring for a leak-resistant seal

For use in hybridization incubators with rotators

35 mm I.D. tubes available in three standard lengths for use in hybridization incubators with rotators. A more convenient method requiring smaller probe solution volumes. Comes with Corning™ standard GL 45 orange plug-seal cap incorporating a silicone O-ring to ensure a leak-resistant seal. The optional Corning vented membrane cap is available for low temperature assays.


Cat. No. 1395-OR is replacement part for silicone o-ring.

Cat. No. 1395-45LTC is replacement part for screw cap with plug seal.

Specifications

Product Type Hybridization Tube
Diameter (English) Inner 1.37 in.
Diameter (Metric) Inner 35 mm
Volume (Metric) 10 mL
Material Borosilicate Glass
Length (Metric) 150 mm
Diameter (Metric) Outer 35 mm
Length (English) 5.90 in.
For Use With (Equipment) Hybridization incubators with rotators
Includes Plug-seal cap, O-ring
Closure Material Polypropylene
Closure Type Plug Seal
Color Orange
Show More Show Less