missing translation for 'onlineSavingsMsg'
Learn More

UVP Hybridization Oven Bottles and Accessories

Supplier:  UVP 07019402

 View more versions of this product

Catalog No. UVP07019402


Only null left

Description

Description

Specifications

Specifications

Polypropylene, PTFE
Hybridization Bottle (30 cm L)
clear
UVP hybridization ovens
285 mL
DNA, RNA, or protein identification
Borosilicate Glass
30 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions