missing translation for 'onlineSavingsMsg'
Learn More

UVP Hybridization Oven Bottles and Accessories

Supplier:  UVP 07019401

 View more versions of this product

Catalog No. UVP07019401


Only null left

Description

Description

Specifications

Specifications

Polypropylene, PTFE
Hybridization Bottle (15 cm L)
clear
UVP hybridization ovens
142.52 mL
DNA, RNA, or protein identification
Borosilicate Glass
15 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions