Learn More
UVP Hybridization Oven Bottles and Accessories
Catalog No. UVP98006703
Change view
Click to view available options
Product Type:
Acrylic Carousel (for upper chamber)
Hybridization Bottle (15 cm L)
Hybridization Bottle (30 cm L)
Rocker Tray
Material:
Borosilicate Glass
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
UVP98006703 Rocker Tray Stainless Steel
UVP07019402 Hybridization Bottle (30 cm L) Borosilicate Glass
UVP76008901 Acrylic Carousel (for upper chamber) Borosilicate Glass
UVP07019401 Hybridization Bottle (15 cm L) Borosilicate Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. UVP98006703 Supplier UVP Supplier No. 98006703
Only null left
Specifications
Product Type Rocker Tray
Material Stainless Steel
For Use With (Application) DNA, RNA, or protein identification
Length (Metric) 31.75 cm
For Use With (Equipment) UVP hybridization ovens