Learn More
Stemcell Technologies Inc ClonaCell™-HY Medium B, Size: 500 mL
Catalog No. NC1728974
Encompass
ClonaCell™-HY Medium B is a serum-free liquid medium used to wash lymphocytes and parental myeloma cells to remove serum prior to fusion and during the fusion process. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Catalog No. NC1728974 Supplier Stemcell Technologies Inc Supplier No. 03802
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ClonaCell™-HY Medium B is a serum-free liquid medium used to wash lymphocytes and parental myeloma cells to remove serum prior to fusion and during the fusion process. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.