Learn More
Andwin Scientific Inc HUMIDITY INDICATOR CARD DFS Item
Supplier:  Andwin Scientific Inc MS200032
The Humidity Indicator Cards are ideal for use as spot indicator. Humidity is less than 30% if the spot is in the "blue" condition. As humidity becomes greater than 30%, the spot will transform from "blue" to "pink"
Catalog No. NC9511648
Only null left
Explore available promotions