missing translation for 'onlineSavingsMsg'
Learn More

Humic acid

CAS: 1415-93-6

$84.90 - $296.58

Chemical Identifiers

CAS 1415-93-6
MDL Number MFCD00147177
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4174706
View Documents
Thermo Scientific Chemicals
04174706
5 g
Each for $84.90
Only null left
 
AA4174714
View Documents
Thermo Scientific Chemicals
04174714
25 g
Each for $296.58
Only null left
 
Description

Description

Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1415-93-6
MFCD00147177

Specifications

1415-93-6
Slightly soluble in water,with much swellingSoluble in organic solvents like acetone and benzenol. Slightly soluble in water.
Crystalline Powder
MFCD00147177
5 g
Humic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-809-6

RTECSNumber : MT6544000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only