missing translation for 'onlineSavingsMsg'
Learn More

Humic acid

Catalog No. AA4174714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4174714 25 g
AA4174706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4174714 Supplier Thermo Scientific Chemicals Supplier No. 04174714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1415-93-6

Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1415-93-6
MDL Number MFCD00147177

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Solubility Information Slightly soluble in water,with much swellingSoluble in organic solvents like acetone and benzenol. Slightly soluble in water.
Chemical Name or Material Humic acid
EINECSNumber 215-809-6
RTECSNumber MT6544000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only