Learn More
Humic acid, sodium salt, 45-70%, technical, (as humic acid)
CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol
$71.69 - $418.94
Chemical Identifiers
| CAS | 68131-04-4 |
|---|---|
| Molecular Formula | C9H8Na2O4 |
| Molecular Weight (g/mol) | 226.139 |
| MDL Number | MFCD00135560 |
| InChI Key | HMGUIQPKFUZDPV-UHFFFAOYSA-L |
| Synonym | humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt |
| PubChem CID | 21908873 |
| IUPAC Name | disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate |
| SMILES | C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC120861000
|
Thermo Scientific Chemicals
120861000 |
100 g | Plastic bottle |
N/A
|
|
||||
|
AC120860010
|
Thermo Scientific Chemicals
120860010 |
1 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 68131-04-4 | |
| 226.139 | |
| HMGUIQPKFUZDPV-UHFFFAOYSA-L | |
| 21908873 | |
| C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+] |
| C9H8Na2O4 | |
| MFCD00135560 | |
| humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt | |
| disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate |
Specifications
| >300.0°C | |
| Black | |
| C9H8Na2O4 | |
| humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt | |
| C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+] | |
| 226.139 | |
| 45 to 70% | |
| 100 g | |
| 14% to 25% (105°C, 4 hrs) | |
| Crystalline Powder |
| 68131-04-4 | |
| 45-70% | |
| MFCD00135560 | |
| HMGUIQPKFUZDPV-UHFFFAOYSA-L | |
| disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate | |
| 21908873 | |
| Technical | |
| Authentic | |
| Plastic bottle | |
| Humic acid, sodium salt, tech., 50-60% (as humic acid) |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 268-608-0
RTECSNumber : MT6550000
TSCA : TSCA
RUO – Research Use Only