missing translation for 'onlineSavingsMsg'
Learn More

Humic acid, sodium salt, 45-70%, technical, (as humic acid)

Catalog No. ac120861000
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120861000 100 g Plastic bottle
AC120860010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120861000 Supplier Thermo Scientific Chemicals Supplier No. 120861000
Only null left

CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68131-04-4
Molecular Formula C9H8Na2O4
Molecular Weight (g/mol) 226.139
MDL Number MFCD00135560
InChI Key HMGUIQPKFUZDPV-UHFFFAOYSA-L
Synonym humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt
PubChem CID 21908873
IUPAC Name disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
SMILES C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color Black
Assay Percent Range 45-70%
Percent Purity 45 to 70%
Grade Technical
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 14% to 25% (105°C, 4 hrs)
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Humic acid, sodium salt, tech., 50-60% (as humic acid)
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 268-608-0
RTECSNumber MT6550000
TSCA TSCA

RUO – Research Use Only