missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HuMEC Basal Serum-Free Medium (1X)
GREENER_CHOICE

Catalog No. 12753018
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12753018 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12753018 Supplier Gibco™ Supplier No. 12753018
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HuMEC Basal Serum-Free Medium (1X)

HuMEC Basal Serum-Free Medium is designed to work with HuMEC Supplement Kit (SKU# 12755013). Together, these products are engineered specifically for the culture of human mammary epithelial cells (SKU# A10565). This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.

Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.

Contents and Storage:
HuMEC Basal Serum Free Medium should be supplemented with HuMEC Supplement Kit. HuMEC Supplement Kit and HuMEC Basal Serum Free Media can be ordered together as HuMEC Ready Medium (SKU# 12752010). The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store the basal medium at +2°C to +8°C.

Specifications

Cell Type Epithelial Cell (Mammary)
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type HuMEC Basal Serum-Free Medium
Sterility Sterile-filtered
With Additives Phenol Red
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.