missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Versican V0, V1 Neo (aa 436-441) Synthetic Peptide

Catalog No. PEP235
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP235 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP235 Supplier Invitrogen™ Supplier No. PEP235

Synthetic Peptide

Human Versican (residues 436-441) Synthetic Peptide, PEP-235, from Invitrogen.

Versican is a member of the family of large aggregating proteoglycans (also including aggrecan, brevican, and neurocan). The protein shows wide tissue distribution which includes fibrous, articular, and elastic cartilages, as well as nervous, epidermal, arterial, and loose connective tissues. Versican V1 has been shown to enhance cell proliferation, and also induce cell transformation and protect cells from apoptosis. This was demonstrated through its ability to downregulate the expression of proapoptotic Bad. Versican's existence as a extracellular matrix protein in human airways has also lead to the discovery of an inflammatory role in asthmatics.

Specifications

Gene ID (Entrez) 1462
Species Human
Purification Method purified
Accession Number P13611
Common Name Human Versican V0, V1 Neo (aa 436-441)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol VCAN
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C G G D(436) P E A A E(441)
Show More Show Less