missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UNC45B (aa 814-927) Control Fragment Recombinant Protein

Catalog No. RP90798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP90798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP90798 Supplier Invitrogen™ Supplier No. RP90798
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53648 (PA5-53648. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

UNC45B plays a role in myoblast fusion and sarcomere organization.

Specifications

Accession Number Q8IWX7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 146862
Name Human UNC45B (aa 814-927) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA445617; cardiomyopathy associated 4; CMYA4; CTRCT43; D230041A13Rik; FLJ38610; MGC119540; MGC119541; Protein unc-45 homolog B; SMUNC45; striated muscle UNC45; UNC45; unc-45 homolog B; unc-45 homolog B (C. elegans); unc-45 myosin chaperone B; Unc45b; UNC-45 B
Common Name UNC45B
Gene Symbol Unc45b
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GEDDDKVQNAAAGALAMLTAAHKKLCLKMTQVTTQWLEILQRLCLHDQLSVQHRGLVIAYNLLAADAELAKKLVESELLEILTVVGKQEPDEKKAEVVQTARECLIKCMDYGFI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less