missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (1 x 106 cells)

Catalog No. C01510C
Encompass
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01510C 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C01510C Supplier Gibco™ Supplier No. C01510C
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (1 x 106 cells)

Primary human umbilical vein endothelial cells, pooled from multiple donors, cryopreserved at the end of the primary culture.
  • Viability at least 70%
  • ≥1,000,000 viable cells/vial
  • Capable of at least 16 population doublings
  • Mycoplasma: Not Detected
  • Hepatitis B: Not Detected
  • Hepatitis C: Not Detected
  • HIV-1: Not Detected
  • Bacteria, yeast, and other fungi: Not Detected
  • Positive for von Willebrand factor (vWf)
  • Positive for CD31
  • Negative for α-actin
  • Positive for diI-Ac-LDL uptake
  • Specifications

    Content And Storage Store at liquid nitrogen temperature.
    Age Newborn (14 days or less)
    Cell Type Endothelial Cells (Umbilical Vein)
    Culture Type Adherent Cell Culture
    Form Cryopreserved
    Species Human
    Donor Source Pooled
    No. of Cells ≥1000000
    Product Line Cascade Biologics
    Product Type Human Umbilical Vein Endothelial Cells
    Quantity 1 vial
    Tested For Hepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
    Show More Show Less
    What types of large vessel endothelial cells do you offer?

    We carry primary human umbilical vein endothelial cells (HUVEC) from one donor or pooled from multiple donors.

    For Research Use Only. Not for use in diagnostic procedures.