Learn More
ACROBiosystems Inc Human TNF-beta / Lymphotoxin-alpha Protein, His Tag (MALS verified)
Numéro de catalogue. 502331360
Encompass
Human TNF-beta Protein, His Tag (TNB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 35 - Leu 205 (Accession # P01374).
Numéro de catalogue. 502331360 Fournisseur ACROBiosystems Inc Code fournisseur TNBH52H3100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Human TNF-beta Protein, His Tag (TNB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 35 - Leu 205 (Accession # P01374).