Learn More
Revvity Health Sciences Human TNF alpha kit, 10000 Tests DFS Item
Supplier:  Perkin Elmer 62HTNFAPEH
TNF alpha is a cytokine mainly produced by macrophages, T cells, and neutrophils. TNF alpha acts as a chemoattractant for neutrophils, and promotes infiltration and activation at inflammation sites. TNF alpha is involved in several cancer types for its ability to promote angiogenesis and cell proliferation, as well as to inhibit apoptosis. Assessment of serum samples often requires enhanced sensitivity. In some cases, AlphaLISA assays may have sufficient sensitivity to enable the detection of low levels of analytes in serum or plasma. Check out the Human TNF AlphaLISA kits analytical performances for more information or learn more about AlphaLISA. When assaying, always follow the recommended protocol and avoid highly haemolyzed samples.
Catalog No. NC1342277
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions