missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMPRSS11D (aa 232-356) Control Fragment Recombinant Protein

Catalog No. RP109646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109646 Supplier Invitrogen™ Supplier No. RP109646
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (60%), Rat (60%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.

Specifications

Accession Number O60235
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9407
Name Human TMPRSS11D (aa 232-356) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Adrenal secretory serine protease; airway spesific trypsin-like protease; airway trypsin like protease; airway trypsin-like protease; AsP; AST; AT; BC020151; HAT; Mat; MGC150587; MGC150588; Rat; TMPRSS11D; Transmembrane protease serine 11 D; Transmembrane protease serine 11 D catalytic chain; Transmembrane protease serine 11 D non-catalytic chain; transmembrane protease, serine 11 D; transmembrane serine protease 11 D
Common Name TMPRSS11D
Gene Symbol TMPRSS11D
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NSNPRDWIATSGISTTFPKLRMRVRNILIHNNYKSATHENDIALVRLENSVTFTKDIHSVCLPAATQNIPPGSTAYVTGWGAQEYAGHTVPELRQGQVRIISNDVCNAPHSYNGAILSGMLCAGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less