Learn More
ACROBiosystems Inc ACROBiosystems Human TIM-1 / HAVCR1 Protein, His Tag(MALSverified)
Catalog No. 502106371
Encompass
Recombinant Protein;100UG;Human TIM-1 / HAVCR1, His Tag (KI1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Gly 295 (Accession # Q96D42-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.5 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 502106371 Supplier ACROBiosystems Inc Supplier No. KI1H52H3100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Human TIM-1 / HAVCR1, His Tag (KI1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Gly 295 (Accession # Q96D42-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.5 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.