Learn More
ACROBiosystems Inc Human TGF-beta RIII / TGFBR3 Protein, His Tag
Catalog No. 502331626
Encompass
Human TGF-beta RIII / TGFBR3 Protein, His Tag (TG3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gly21-Val787 (Accession # Q03167-1).
Catalog No. 502331626 Supplier ACROBiosystems Inc Supplier No. TG3H52H3100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Human TGF-beta RIII / TGFBR3 Protein, His Tag (TG3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gly21-Val787 (Accession # Q03167-1).