Learn More
ACROBiosystems Inc Human Tau-441 (P301L), His Tag (TAU-H5144) is expressed from E.coli cells. It contains AA Met 1 - Leu 441 (Accession # P10636-8 (P301L)).
Catalog No. 502253959
Encompass
Tau is a microtubule-associated protein, which encodes by the MAPT gene that located on chromosome 17q21. Tau Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. Hyperphosphorylation of the tau protein (tau inclusions, pTau) can result in the self-assembly of tangles of paired helical filaments and straight filaments, which are involved in the pathogenesis of Alzheimer's disease, frontotemporal dementia, and other tauopathies.
Catalog No. 502253959 Supplier ACROBiosystems Inc Supplier No. TAUH5144100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tau is a microtubule-associated protein, which encodes by the MAPT gene that located on chromosome 17q21. Tau Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. Hyperphosphorylation of the tau protein (tau inclusions, pTau) can result in the self-assembly of tangles of paired helical filaments and straight filaments, which are involved in the pathogenesis of Alzheimer's disease, frontotemporal dementia, and other tauopathies.