missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Suspension Hepatocytes, Metabolism Qualified, Male, (4-8 x 106 cells)
Description
These cryopreserved human male hepatocytes are suitable for use in suspension metabolism applications.
Cells per vial: 4-8 million
The commonly tested Phase I (P450) and Phase II (UGT, SULT) drug metabolizing enzymes were analyzed for metabolic activity in each lot. Metabolite formation was measured by standard biochemical assays using LC⁄MS⁄MS or HPLC.
Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
Phase I and II enzyme activities
Proper morphology
Viabilities generally >75%
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Learn more about our cryopreserved hepatocytes
Learn about our full line of hepatocytes ,
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Cells per vial: 4-8 million
The commonly tested Phase I (P450) and Phase II (UGT, SULT) drug metabolizing enzymes were analyzed for metabolic activity in each lot. Metabolite formation was measured by standard biochemical assays using LC⁄MS⁄MS or HPLC.
Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Learn more about our cryopreserved hepatocytes
Learn about our full line of hepatocytes ,
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Specifications
Specifications
| Content And Storage | Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer. |
| Cell Type | Hepatocytes |
| Culture Type | Suspension Cell Culture |
| Form | Cryopreserved |
| Gender | Male |
| Species | Human |
| Donor Source | Single Donor |
| No. of Cells | 4-8 x 106 |
| Product Line | Gibco |
| Product Type | Hepatocytes |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.