Learn More
Biomatik Corporation Human sTfR1 (Soluble Transferrin Receptor1) ELISA Kit, 96 tests
Catalog No. 501500793
Encompass
Human sTfR1 (Soluble Transferrin Receptor1) ELISA Kit for detecting sTfR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000ng/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02786. Target Synonyms: sTfR1; Soluble Transferrin Receptor1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501500793 Supplier Biomatik Corporation Supplier No. EKF57147
Human sTfR1 (Soluble Transferrin Receptor1) ELISA Kit for detecting sTfR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000ng/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02786. Target Synonyms: sTfR1; Soluble Transferrin Receptor1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.