Human SP-A1 (Pulmonary surfactant-associated protein A1) ELISA Kit for detecting SP-A1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q8IWL2. Target Synonyms: MGC133365; PSAP; PSPA; SFTP1; SFTPA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.