missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SMKR1 (aa 1-65) Control Fragment Recombinant Protein

Catalog No. RP108142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108142 Supplier Invitrogen™ Supplier No. RP108142
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (30%), Rat (30%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67493 (PA5-67493. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SMKR1 is a protein coding gene.

Specifications

Accession Number H3BMG3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 100287482
Name Human SMKR1 (aa 1-65) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias small lysine rich protein 1; small lysine-rich protein 1; SMKR1
Common Name SMKR1
Gene Symbol SMKR1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPAKGKKGKGQGKSHGKKQKKPEVDILSPAAMLNLYYIAHNVADCLHLRGFHWPGAPKGKKGRSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less